Learn More
Strem, An Ascensus Company CAS# 14075-53-7. 500g. Potassium tetrafluoroborate. MFCD00011395
SDP
Catalog No. 5090114301
Encompass
CAS# 14075-53-7. 500g. Potassium tetrafluoroborate. MFCD00011395. Molecular Weight: 125.91. Molecular Formula: KBF4. Color/form: white pwdr. Strem# 93-1951. http://www.strem.com/catalog/v/93-1951/
Catalog No. 50-901-14301 Supplier Strem, An Ascensus Company Supplier No. 931951500G
CAS# 14075-53-7. 500g. Potassium tetrafluoroborate. MFCD00011395. Molecular Weight: 125.91. Molecular Formula: KBF4. Color/form: white pwdr. Strem# 93-1951. http://www.strem.com/catalog/v/93-1951/