Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tetrafluoroborate, 99%

Catalog No. p-7048820
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1155636 500 g
AA11556A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1155636 Supplier Thermo Scientific Chemicals Supplier No. 01155636
Only null left
Add to Cart

CAS: 14075-53-7 | BF4K | 125.90 g/mol

Potassium tetrafluoroborate is used as an active filler in the preparation of resin-bonded abrasives. It is also used for extraction, refining and processing of metals in the chemical industry. It finds application as an additive in the alloying industry and welding agent. Further, it is used in the production of fluxing agents for brazing and soldering.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14075-53-7
Molecular Formula BF4K
Molecular Weight (g/mol) 125.90
MDL Number MFCD00011395
InChI Key AKEBROIVCDHVSD-UHFFFAOYSA-N
Synonym potassium tetrafluoroborate, potassium fluoroborate, potassium fluoborate, potassium borofluoride, avogadrite, potassium boron fluoride, potassium fluorohydroborate, potassium hydrofluoroborate, boron potassium fluoride, potassium boride fluoride
PubChem CID 518872
IUPAC Name potassium;tetrafluoroborate
SMILES [K+].F[B-](F)(F)F

Specifications

Boiling Point (decomposition)
Melting Point 529°C
Physical Form Powder
Quantity 500 g
Linear Formula KBF4
Merck Index 14,7686
Solubility Information Soluble in water (0;45g/100g H2O); Slightly soluble in boiling alcoholSlightly soluble in water and hot ethyl alcohol; Insoluble in cold ethyl alcohol.
Formula Weight 125.91
Percent Purity ≤99%
Refractive Index 1.3245
Sensitivity Hygroscopic
Density 2.50 g/mL
Assay Typically >99.5%
Chemical Name or Material Potassium tetrafluoroborate
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
EINECSNumber 237-928-2
RTECSNumber ED2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.