Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium tetrathionate

Catalog No. 503126113 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-6113 25 g
50-312-6112 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-312-6113 Supplier Sigma Aldrich Supplier No. P292625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The crystal structure of potassium tetrathionate is monoclinic with a=22.081o, b=7.991o, c=10.108oUsed in the preparation of tetrathionate hydrolase assay from Acidithiobacillus caldus, it acts as an energy source.

Specifications

CAS 13932-13-3
Density 2.96 g/mL (at 25°C)
Molecular Formula K2O6S4
Linear Formula KOSO2SSSO3K
Quantity 25 g
MDL Number MFCD00079233
Synonym Tetrathionic acid dipotassium salt
Molecular Weight (g/mol) 302.45
Percent Purity ≥98.0% (titration)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.