Learn More
Selleck Chemical LLC Potassium thioacetate
SDP
Catalog No. 5000004266
Encompass_Preferred
Potassium thioacetate is widely used as a sulfur source in the synthesis of sulfur-containing organic compounds It has been employed for the synthesis of heterocycles polymers transition-metal ligands nanoparticles bioactive compounds and macromolecular inclusion complexes
Catalog No. 50-000-04266 Supplier Selleck Chemical LLC Supplier No. S602025MG
Potassium thioacetate is widely used as a sulfur source in the synthesis of sulfur-containing organic compounds It has been employed for the synthesis of heterocycles polymers transition-metal ligands nanoparticles bioactive compounds and macromolecular inclusion complexes