Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Thiocyanate, 0.0250 N (N/40), Ricca Chemical
SDP

Catalog No. R6564000500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R6564000500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R6564000500 Supplier Ricca Chemical Company Supplier No. R6564000500A
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 333-20-0
Molecular Weight (g/mol) 97.18 g/mol
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

pH Neutral
Physical Form Solution
Quantity 500 mL
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Odor Odorless
Packaging Natural Poly Bottle
Appearance Clear colorless liquid
Chemical Name or Material Potassium Thiocyanate
Recommended Storage General
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.