Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Thiocyanate, 0.100 N (N/10), Ricca Chemical
SDP

Catalog No. 65805 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6580-5 20 L Cubitainer
658016 500 mL Natural Poly Bottle
6580-32 1 L Natural Poly Bottle
6580-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6580-5 Supplier Ricca Chemical Company Supplier No. 65805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 333-20-0 , 7732-18-5
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Color Colorless
CAS Min % 97.0, 0.97
CAS Max % 100.0, 1.01
Grade Laboratory
Packaging Cubitainer
pH Neutral
Quantity 20 L
Physical Form Liquid
Concentration or Composition Notes 0.100N
Chemical Name or Material Potassium Thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.