Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Thiocyanate, 10% (w/v), Ricca Chemical
SDP

Catalog No. 655532 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6555-32 1 L Natural Poly Bottle
6555-16 500 mL Natural Poly Bottle
6555-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6555-32 Supplier Ricca Chemical Company Supplier No. 655532
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Percent Purity 10%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
pH Neutral
Quantity 1 L
Chemical Name or Material Potassium Thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.