Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium thiocyanate ACS2.5KG
Catalog No. 502897651
Encompass
Potassium thiocyanate is an inorganic potassium salt. Phase transitions in potassium thiocyanate (KSCN) have been investigated by X-ray diffraction studies. The order-disorder type transition with respect to the orientation of the thiocyanate ions was identified. Its polarized infrared spectrum has been reported. It participates in the ring-opening of aziridines. The reaction was carried out in the presence of sulfated zirconia to afford the corresponding -aminothiocyanates.
Catalog No. 50-289-7651 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2077992.5KG
Potassium thiocyanate is an inorganic potassium salt. Phase transitions in potassium thiocyanate (KSCN) have been investigated by X-ray diffraction studies. The order-disorder type transition with respect to the orientation of the thiocyanate ions was identified. Its polarized infrared spectrum has been reported. It participates in the ring-opening of aziridines. The reaction was carried out in the presence of sulfated zirconia to afford the corresponding -aminothiocyanates.