Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Potassium Thiocyanate 10% w/v Aqueous Sol. (1L)
SDP

Catalog No. NC9784874
Encompass

Potassium Thiocyanate 10% w/v Aqueous Sol. (1L)

Catalog No. NC9784874 Supplier Aqua Solutions Supplier No. ACB068
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Thiocyanate 10% w/v Aqueous Sol. (1L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.