Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Thioglycollate, 40% Soln, MP Biomedicals

Catalog No. ICN218163 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN218163 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN218163 Supplier MP Biomedicals, Inc Supplier No. 0521816380

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34452-51-2
Molecular Formula C2H3KO2S
Molecular Weight (g/mol) 130.202
InChI Key HYTYHTSMCRDHIM-UHFFFAOYSA-M
Synonym potassium thioglycolate, unii-xp5wzo5w6h, potassium 2-mercaptoacetate, acetic acid, mercapto-, monopotassium salt, xp5wzo5w6h, 68-11-1 parent, potassium sulfanylacetate, potassium mercaptoacetate, kopcolate, potassium ion thioglycolate
PubChem CID 23688957
IUPAC Name potassium;2-sulfanylacetate
SMILES C(C(=O)[O-])S.[K+]

Specifications

Formula Weight 130.21
Quantity 100 g
Chemical Name or Material Potassium Thioglycollate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.