Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Titanium Oxide Oxalate Dihydrate

Catalog No. 140076X1KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 kg
6 x 1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-007-6X1KG 6 x 1 kg
14-007-1KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-007-6X1KG Supplier Sigma Aldrich Supplier No. 140076X1KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium titanium oxide oxalate dihydrate can be used as a precursor to synthesize titanium dioxide (TiO2) nanoparticles.

Specifications

CAS 14402-67-6
Molecular Formula C4K2O9Ti -+ 2H2O
Linear Formula C4K2O9Ti -+ 2H2O
MDL Number MFCD00150592
Quantity 6 x 1 kg
Molecular Weight (g/mol) 354.13 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.