Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium titanium oxide

Catalog No. AAB2026322
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2026322 100 g
AAB2026336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2026322 Supplier Thermo Scientific Chemicals Supplier No. B2026322
Add to Cart

CAS: 12030-97-6 | K2O3Ti | 174.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12030-97-6
Molecular Formula K2O3Ti
Molecular Weight (g/mol) 174.06
MDL Number MFCD00058803
InChI Key KEWYKJOOCFOYTD-UHFFFAOYSA-N
Synonym kaliumtitanat, titanate,dipotassium, potassiumtitanatewhisker, 2k.o3ti, titanate tio32-,dipotassium, dipotassium;dioxido oxo titanium
PubChem CID 45052161
SMILES [O--].[O--].[O--].[K+].[K+].[Ti+4]

Specifications

Melting Point 1615°C
Quantity 100 g
IUPAC Name dipotassium;dioxido(oxo)titanium
Odor Odorless
Chemical Name or Material Potassium titanium oxide
EINECSNumber 234-748-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.