Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tricyanomethanide

Catalog No. p-7049740
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2070203 1 g
AA2070206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2070203 Supplier Thermo Scientific Chemicals Supplier No. 02070203
Only null left
Add to Cart

CAS: 34171-69-2 | C4KN3 | 129.163 g/mol

Potassium tricyanomethanide is a precursor used for the preparation of dicyano ketene aminals. It reacts with amines to prepare alkyl ammonium tricyanomethanide. Further, it is involved in the preparation of tricyanomethanide-bridged binuclear organometallic complexes such as Cp(dppe)FeC(CN)3 and Cp(PPh3)2RuC(CN)3. It is used as starting material utilized in the preparation of 5-Amino-3-bromo-1-(tert-butyl)-1H-pyrazole-4-carbonitrile, which undergoes sandmeyer reaction to get the corresponding diaminopyrazole, 5-amino-3-aryl-1-(tert-butyl)-1H-pyrazole-4-carboxamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34171-69-2
Molecular Formula C4KN3
Molecular Weight (g/mol) 129.163
MDL Number MFCD00058850
InChI Key ZKJPYQKGNUBNOA-UHFFFAOYSA-N
Synonym potassium tricyanomethanide, potassium methanetricarbonitrile, potassium cyanoform, acmc-1ajhe, potassium ion tricyanomethanide, tricyanomethanide potassium salt, potassiomethanetricarbonitrile
PubChem CID 10964525
IUPAC Name potassium;methanetricarbonitrile
SMILES C(#N)[C-](C#N)C#N.[K+]

Specifications

Color White
Linear Formula KC(CN)3
Quantity 1 g
UN Number UN3439
Solubility Information Soluble in water.
Formula Weight 129.16
Physical Form Crystalline
Chemical Name or Material Potassium tricyanomethanide
Hazard Category H301+H311-H330
Hazard Statement GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.