Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Triethylborohydride Solution

Catalog No. 213438100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-343-8100ML 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 21-343-8100ML Supplier Sigma Aldrich Supplier No. 213438100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reducing agent for synthesis of PtNi intermetallic nanoparticle catalysts with electrocatalytic activityReducing agent for preparation of vanadium nanoparticles (~7 nm) stabilized on activated carbonReagent for:Preparation of lithium, sodium, and potassium boron monohydride complexes with tetramethylethylenediamine and pentamethyldiethylenetriamineDehydrogenation reactionsTandem nucleophilic addition/C-C bond cleaving fragmentation reactions with cyclic vinylogous acyl triflatesPreparation of coordinatively unsaturated pseudotetrahedral iron(II) precursors as precatalysts for the room-temperature hydrogenation of unsaturated hydrocarbonsSubstitution reactions

Specifications

CAS 7704-99-6
Molecular Formula H2Zr
Linear Formula ZrH2
MDL Number MFCD00148878
Quantity 100 g
Synonym Potassium hydrotriethylborate; Potassium triethylhydridoborate
Molecular Weight (g/mol) 93.24 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.