Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium trifluoromethanesulfonate

Catalog No. 4228435G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-284-35G 5 g
42-284-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42-284-35G Supplier Sigma Aldrich Supplier No. 4228435G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Potassium trifluoromethanesulfonate (kotf, potassium triflate) is the potassium salt of trifluoromethanesulphonic acid. it has been prepared by neutralizing a warm aqueous solution of trifluoromethanesulphonic acid with potassium carbonate. the structure of siloxane-poly(oxyethylene) hybrids doped with potassium triflate has been investigated.potassium trifluoromethanesulfonate (potassium triflate) is suitable as a reagent in the synthesis of guanine-quadruplex hybrid materials by the self-assembly of a guanine-siloxane monomer.it may be used in the following studies:
  • As a template source for constructing the heterotopic isothiosemicarbazide-based macrocyclic ligand
  • As a supporting electrolyte in the electrochemical study of evidence for gold anion in ethylenediamine
  • As a reagent in the synthesis of imidazolium salt, 3-methyl-1-(3r,3ar,6s,6ar)-[6-(benzyloxy)-hexahydrofuro[3,2-b]furan-3-yl]imidazolium trifluoromethanesulfonate

Specifications

CAS 2926-27-4
Molecular Formula CF3KO3S
Linear Formula CF3SO3K
MDL Number MFCD00042370
Quantity 5 g
Synonym Potassium triflate; Trifluoromethanesulfonic acid potassium salt
Molecular Weight (g/mol) 188.17
Percent Purity 98%
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.