Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Trimethylsilanolate

Catalog No. 32486825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-486-825G 25 g
32-486-81KG 1 kg
32-486-8100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 32-486-825G Supplier Sigma Aldrich Supplier No. 32486825G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium Trimethylsilanolate

Specifications

CAS 10519-96-7
Melting Point 135°C to 138°C
Molecular Formula C3H9KOSi
Linear Formula (CH3)3SiOK
MDL Number MFCD00013421
Quantity 25 g
Synonym Trimethylsilanol potassium salt
Molecular Weight (g/mol) 128.29 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.