Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tungstate, 99.95%, for trace metal analysis, -100 mesh

Catalog No. AC448330050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC448330050 5 g Glass bottle
AC448330250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC448330050 Supplier Thermo Scientific Chemicals Supplier No. 448330050
Add to Cart

CAS: 7790-60-5 | K2O4W | 326.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-60-5
Molecular Formula K2O4W
Molecular Weight (g/mol) 326.05
InChI Key AAQNGTNRWPXMPB-UHFFFAOYSA-N
Synonym potassium tungstate, unii-t6vrm0ou6v, t6vrm0ou6v, dipotassium dioxido dioxo tungsten, dipotassium wolframate, potassium tungstate vi, tungstate wo42-, dipotassium, t-4, 2k.wo4, potassium tungstate-w
PubChem CID 3084032
IUPAC Name dipotassium;dioxido(dioxo)tungsten
SMILES [O-][W](=O)(=O)[O-].[K+].[K+]

Specifications

Melting Point 921°C
Quantity 5 g
Assay Percent Range 99.95%
Linear Formula K2WO4
Merck Index 15,7811
Formula Weight 326.05
Percent Purity 99.95%
Grade Trace Metal
Packaging Glass bottle
Chemical Name or Material Potassium tungstate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 232-215-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.