Learn More
Bioworld POTASSM PHOSPHATE BUFFR 0.1,1L
Catalog No. NC2226617
Encompass_Preferred
Potassium Phosphate Buffer 0.1M, pH 7.4
This item is not returnable. View return policy
Catalog No. NC2226617 Supplier Bioworld Supplier No. 416201642
Potassium Phosphate Buffer 0.1M, pH 7.4