Learn More
IBI SCIENTIFIC 3P Potato Dextrose Agar, 500 grams
Catalog No. NC0753549
Encompass_Preferred
Potato Dextrose Agar - 500gm Potato Dextrose Agar is recommended for the cultivation and isolation of yeasts and molds from dairy and food products. By lowering the pH to 3.5 +/- 0.1 inhibits bacterial growth and aids in the isolation of fungi.
Catalog No. NC0753549 Supplier IBI SCIENTIFIC 3P Supplier No. IB49190
Potato Dextrose Agar - 500gm Potato Dextrose Agar is recommended for the cultivation and isolation of yeasts and molds from dairy and food products. By lowering the pH to 3.5 +/- 0.1 inhibits bacterial growth and aids in the isolation of fungi.