Learn More
Alkali Scientific Potato Dextrose Agar, 1 Kg
SDP
Catalog No. 5000660655
Encompass_Preferred
Potato Dextrose Agar (PDA), 1kg, is a widely used medium for cultivating fungi, especially yeasts and molds, from various environmental, food, and clinical samples. This medium contains dextrose as a primary carbon source and potato infusion, which provides essential vitamins, amino acids, and other nutrients required for fungal growth. PDA is particularly effective in supporting the growth of a wide range of fungal species, making it a standard medium in mycology laboratories. The 1kg pack size is ideal for high-throughput laboratories or for use in fungal isolation, environmental testing, and microbiological analysis.
Catalog No. 50-006-60655 Supplier Alkali Scientific Supplier No. ATO001CP
Potato Dextrose Agar (PDA), 1kg, is a widely used medium for cultivating fungi, especially yeasts and molds, from various environmental, food, and clinical samples. This medium contains dextrose as a primary carbon source and potato infusion, which provides essential vitamins, amino acids, and other nutrients required for fungal growth. PDA is particularly effective in supporting the growth of a wide range of fungal species, making it a standard medium in mycology laboratories. The 1kg pack size is ideal for high-throughput laboratories or for use in fungal isolation, environmental testing, and microbiological analysis.