Learn More
Alkali Scientific Potato Dextrose Agar, 10 Kg
SDP
Catalog No. 5000660441
Encompass_Preferred
Potato Dextrose Agar (PDA), 10kg, is a large-volume formulation designed for growing fungi from a wide range of environmental, clinical, and food samples. The medium is rich in dextrose and potato infusion, which provide essential nutrients to support the growth of yeasts and molds. This 10kg pack size is ideal for high-volume laboratories, pharmaceutical companies, or food safety testing facilities that require large quantities of PDA for fungal isolation and environmental monitoring. It is also suitable for research applications where extensive fungal cultures are required for experiments, strain isolation, or microbial assays.
Catalog No. 50-006-60441 Supplier Alkali Scientific Supplier No. ATO004CP
Potato Dextrose Agar (PDA), 10kg, is a large-volume formulation designed for growing fungi from a wide range of environmental, clinical, and food samples. The medium is rich in dextrose and potato infusion, which provide essential nutrients to support the growth of yeasts and molds. This 10kg pack size is ideal for high-volume laboratories, pharmaceutical companies, or food safety testing facilities that require large quantities of PDA for fungal isolation and environmental monitoring. It is also suitable for research applications where extensive fungal cultures are required for experiments, strain isolation, or microbial assays.