Learn More
Bioworld Potato Dextrose Agar, 500 g
Catalog No. 50254477
Encompass_Preferred
Potato Dextrose AgarUsed for the cultivation and enumeration of yeasts and molds especially Fusarium speciesTo inhibit bacterial growth, the medium may be acidified to pH 3.5 by adding tartaric acid, lactic acid or some other organic acid The medium must not be heated after the addition of the acid, this will result in the hydrolysis of the agar and destroy its gelling propertiespH: 5.6Dehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light amber agarSoluble in boiling waterComposition (g/L):Potato Infusio- 4g Glucose-20gAgar - 15g
Catalog No. 50-254-477 Supplier Bioworld Supplier No. 306260051
Potato Dextrose AgarUsed for the cultivation and enumeration of yeasts and molds especially Fusarium speciesTo inhibit bacterial growth, the medium may be acidified to pH 3.5 by adding tartaric acid, lactic acid or some other organic acid The medium must not be heated after the addition of the acid, this will result in the hydrolysis of the agar and destroy its gelling propertiespH: 5.6Dehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light amber agarSoluble in boiling waterComposition (g/L):Potato Infusio- 4g Glucose-20gAgar - 15g