Learn More
Alkali Scientific Potato Dextrose Agar, 15x100mm, 25mL, 10/pk
SDP
Catalog No. 5000662892
Encompass_Preferred
Potato Dextrose Agar (PDA), 15×100mm plates with 25 mL of medium per plate, is a general purpose culture medium widely used for the cultivation, isolation, and enumeration of yeasts and molds. The formulation contains potato infusion and dextrose, providing a rich carbohydrate source that supports robust fungal growth while encouraging sporulation and pigment development in many mold species. PDA is commonly used in clinical mycology, food and beverage quality testing, pharmaceutical quality control, and environmental microbiology laboratories. The nutrient rich composition makes it suitable for maintaining fungal stock cultures as well as routine microbial monitoring. Each plate is pre poured with sterile agar to a consistent depth, ensuring reliable growth conditions, reproducible results, and convenient handling in laboratory workflows.
Catalog No. 50-006-62892 Supplier Alkali Scientific Supplier No. PDAP01
Potato Dextrose Agar (PDA), 15×100mm plates with 25 mL of medium per plate, is a general purpose culture medium widely used for the cultivation, isolation, and enumeration of yeasts and molds. The formulation contains potato infusion and dextrose, providing a rich carbohydrate source that supports robust fungal growth while encouraging sporulation and pigment development in many mold species. PDA is commonly used in clinical mycology, food and beverage quality testing, pharmaceutical quality control, and environmental microbiology laboratories. The nutrient rich composition makes it suitable for maintaining fungal stock cultures as well as routine microbial monitoring. Each plate is pre poured with sterile agar to a consistent depth, ensuring reliable growth conditions, reproducible results, and convenient handling in laboratory workflows.