Learn More
Alkali Scientific Potato Dextrose Agar, 5 Kg
SDP
Catalog No. 5000660761
Encompass_Preferred
Potato Dextrose Agar (PDA), 5kg, is a highly effective medium for growing a wide variety of fungi, including yeasts and molds. It is particularly useful in environmental monitoring, food testing, and clinical diagnostics, where fungal isolation is needed. This medium contains dextrose as the primary carbon source and potato infusion for added nutrients, making it ideal for cultivating fungi from diverse samples. The 5kg bulk size is perfect for high-throughput laboratories or those involved in large-scale testing, offering a cost-effective solution for routine fungal cultivation, research, and microbiological analysis.
Catalog No. 50-006-60761 Supplier Alkali Scientific Supplier No. ATO003CP
Potato Dextrose Agar (PDA), 5kg, is a highly effective medium for growing a wide variety of fungi, including yeasts and molds. It is particularly useful in environmental monitoring, food testing, and clinical diagnostics, where fungal isolation is needed. This medium contains dextrose as the primary carbon source and potato infusion for added nutrients, making it ideal for cultivating fungi from diverse samples. The 5kg bulk size is perfect for high-throughput laboratories or those involved in large-scale testing, offering a cost-effective solution for routine fungal cultivation, research, and microbiological analysis.