Learn More
Alkali Scientific Potato Dextrose Agar, 500 G
SDP
Catalog No. 5000660859
Encompass_Preferred
Potato Dextrose Agar (PDA), 500g, is a versatile and commonly used fungal growth medium in microbiology. The medium contains dextrose, which serves as an energy source, and potato infusion, which provides nutrients necessary for fungal growth. This formulation is specifically designed to support the cultivation of a wide variety of fungi, including yeasts and molds, from clinical, environmental, and food samples. The 500g pack size is ideal for laboratories that require a moderate volume of fungal growth medium for regular fungal isolation, contamination testing, and environmental monitoring.
Catalog No. 50-006-60859 Supplier Alkali Scientific Supplier No. ATO002CP
Potato Dextrose Agar (PDA), 500g, is a versatile and commonly used fungal growth medium in microbiology. The medium contains dextrose, which serves as an energy source, and potato infusion, which provides nutrients necessary for fungal growth. This formulation is specifically designed to support the cultivation of a wide variety of fungi, including yeasts and molds, from clinical, environmental, and food samples. The 500g pack size is ideal for laboratories that require a moderate volume of fungal growth medium for regular fungal isolation, contamination testing, and environmental monitoring.