Learn More
HIMEDIA Potato Dextrose Agar, Granulated, GM096, Microbiology Culture Media, Dehydrated Culture Media, Granulated Agar, Yeast and Mould Enumeration, Low Dust, Powder, 500 g
Catalog No. NC4013624
Encompass_Preferred
Potato Dextrose Agar, Granulated is a classic mycological medium for isolating and enumerating yeasts and moulds from water, dairy, other foods and clinical samples. Potato infusion and dextrose encourage luxuriant fungal growth and sporulation while agar solidifies the medium at final pH 5.6. Acidification with sterile tartaric acid suppresses bacteria when required. The granulated form is low-dusting, flows freely and rehydrates rapidly for easier, safer preparation. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013624 Supplier HIMEDIA Supplier No. GM096500G
Potato Dextrose Agar, Granulated is a classic mycological medium for isolating and enumerating yeasts and moulds from water, dairy, other foods and clinical samples. Potato infusion and dextrose encourage luxuriant fungal growth and sporulation while agar solidifies the medium at final pH 5.6. Acidification with sterile tartaric acid suppresses bacteria when required. The granulated form is low-dusting, flows freely and rehydrates rapidly for easier, safer preparation. Dehydrated powder, store in a cool dry place. Packaged as 500 g.