Learn More
Alkali Scientific Potato Dextrose Agar (PDA) Deep Fill 15mL Tube, 10/Pk
SDP
Catalog No. 5000663132
Encompass_Preferred
Potato Dextrose Agar (PDA) is a widely used general purpose medium for the cultivation, isolation, and enumeration of yeasts and molds. It is commonly used in clinical, food, and cosmetic microbiology laboratories for fungal testing and environmental monitoring. This product features a 15 mL deep fill tube format that provides significantly more medium than standard slants. The larger volume allows the agar to be melted and poured into petri dishes when fresh plates are required, or solidified as a deep or slant for long term fungal culture storage. The formulation includes potato infusion as a nutrient base that stimulates spore formation and pigment development, while dextrose supplies a readily available carbohydrate source that supports robust fungal growth.
Catalog No. 50-006-63132 Supplier Alkali Scientific Supplier No. PDAT00
Potato Dextrose Agar (PDA) is a widely used general purpose medium for the cultivation, isolation, and enumeration of yeasts and molds. It is commonly used in clinical, food, and cosmetic microbiology laboratories for fungal testing and environmental monitoring. This product features a 15 mL deep fill tube format that provides significantly more medium than standard slants. The larger volume allows the agar to be melted and poured into petri dishes when fresh plates are required, or solidified as a deep or slant for long term fungal culture storage. The formulation includes potato infusion as a nutrient base that stimulates spore formation and pigment development, while dextrose supplies a readily available carbohydrate source that supports robust fungal growth.