Learn More
Alkali Scientific Potato Dextrose Agar (PDA) w/o Tartaric Acid 25mL Plate, 10/Pk
SDP
Catalog No. 5000662891
Encompass_Preferred
Potato Dextrose Agar (PDA) without tartaric acid is a general purpose fungal culture medium used for the cultivation and enumeration of yeasts and molds. Unlike acidified PDA formulations, this medium retains the standard pH of approximately 5.6, allowing a wider range of fungi to grow while maintaining the classic PDA nutrient composition. The formulation includes potato infusion for rich nutrients that encourage sporulation and pigment development, while dextrose provides an energy source for fungal metabolism. This product is poured as a 25 mL deep fill plate, providing a larger moisture reservoir than standard plates. The additional depth helps prevent agar drying and cracking during extended incubation periods commonly required for slow growing fungal organisms.
Catalog No. 50-006-62891 Supplier Alkali Scientific Supplier No. PDAP03
Potato Dextrose Agar (PDA) without tartaric acid is a general purpose fungal culture medium used for the cultivation and enumeration of yeasts and molds. Unlike acidified PDA formulations, this medium retains the standard pH of approximately 5.6, allowing a wider range of fungi to grow while maintaining the classic PDA nutrient composition. The formulation includes potato infusion for rich nutrients that encourage sporulation and pigment development, while dextrose provides an energy source for fungal metabolism. This product is poured as a 25 mL deep fill plate, providing a larger moisture reservoir than standard plates. The additional depth helps prevent agar drying and cracking during extended incubation periods commonly required for slow growing fungal organisms.