Learn More
Bioworld Potato Dextrose Broth, 500 g
Catalog No. 50254478
Encompass_Preferred
Potato Dextrose BrothUsed for the cultivation of yeasts and molds especially Fusarium speciesTo inhibit bacterial growth, the medium may be acidified to pH 3.5 by adding tartaric acid, lactic acid or some other organic acid The medium must not be heated after the addition of the acid, this will result in the hydrolysis of the agar and destroy its gelling propertiesPrepared Appearance: light amber brothSoluble in boiling waterComposition:Potato Infusion Glucose
Catalog No. 50-254-478 Supplier Bioworld Supplier No. 306260061
Potato Dextrose BrothUsed for the cultivation of yeasts and molds especially Fusarium speciesTo inhibit bacterial growth, the medium may be acidified to pH 3.5 by adding tartaric acid, lactic acid or some other organic acid The medium must not be heated after the addition of the acid, this will result in the hydrolysis of the agar and destroy its gelling propertiesPrepared Appearance: light amber brothSoluble in boiling waterComposition:Potato Infusion Glucose