Learn More
NEOGEN Potato Dextrose Broth, 500 g
SDP
Catalog No. 502015606
Encompass_Preferred
Potato Dextrose Broth is used for the cultivation of fungi in a laboratory setting. Potato Dextrose Broth is not intended for use in the diagnosis of disease or other conditions in humans. Potato Dextrose Broth is a general purpose broth for yeasts and molds. The low pH of this medium inhibits bacterial growth. Potato Dextrose Broth is the same formula as Potato Dextrose Agar, but agar has been omitted.For more information visit https://www.neogen.com/solutions/Microbiology/potato-dextrose-broth
Catalog No. 50-201-5606 Supplier NEOGEN Supplier No. 700003424
Potato Dextrose Broth is used for the cultivation of fungi in a laboratory setting. Potato Dextrose Broth is not intended for use in the diagnosis of disease or other conditions in humans. Potato Dextrose Broth is a general purpose broth for yeasts and molds. The low pH of this medium inhibits bacterial growth. Potato Dextrose Broth is the same formula as Potato Dextrose Agar, but agar has been omitted.For more information visit https://www.neogen.com/solutions/Microbiology/potato-dextrose-broth