Learn More
HIMEDIA Potato Dextrose Broth, Granulated, GM403, Microbiology Culture Media, Dehydrated Culture Media, Granulated Broth, Yeast and Mould Cultivation, Low Dust, Powder, 500 g
Catalog No. NC4013626
Encompass_Preferred
Potato Dextrose Broth, Granulated is a liquid mycological medium for isolating, cultivating and enumerating yeasts and moulds. Potato infusion and dextrose drive luxuriant fungal growth at final pH 5.1, making it well suited to enrichment, spoilage investigations and biomass production in food, water and clinical work. The granulated form is low-dusting, flows freely and rehydrates quickly for safer, more consistent preparation. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013626 Supplier HIMEDIA Supplier No. GM403500G
Potato Dextrose Broth, Granulated is a liquid mycological medium for isolating, cultivating and enumerating yeasts and moulds. Potato infusion and dextrose drive luxuriant fungal growth at final pH 5.1, making it well suited to enrichment, spoilage investigations and biomass production in food, water and clinical work. The granulated form is low-dusting, flows freely and rehydrates quickly for safer, more consistent preparation. Dehydrated powder, store in a cool dry place. Packaged as 500 g.