Learn More
Bioworld Potato Infusion Agar, 500 g
Catalog No. 50254355
Encompass_Preferred
Potato Infusion AgarFor the isolation of Brucella abortus, especially in mass cultivation procedures pH: 6.8Working Concentration: 49 g/LDehydrated Appearance: medium tan, homogeneous powderPrepared Appearance: medium amber agarSoluble in boiling water containing 2% glycerolContains:Potatoes, Infusion From Agar Glucose Proteose Peptone Beef Extract Sodium Chloride Glycerol
Catalog No. 50-254-355 Supplier Bioworld Supplier No. 306200661
Potato Infusion AgarFor the isolation of Brucella abortus, especially in mass cultivation procedures pH: 6.8Working Concentration: 49 g/LDehydrated Appearance: medium tan, homogeneous powderPrepared Appearance: medium amber agarSoluble in boiling water containing 2% glycerolContains:Potatoes, Infusion From Agar Glucose Proteose Peptone Beef Extract Sodium Chloride Glycerol