Learn More
Alkali Scientific Potato Starch, Insoluble, 100 Grams
SDP
Catalog No. 5000660870
Encompass_Preferred
Potato Starch, Insoluble, is a high-purity starch used in laboratory applications requiring a stable, insoluble carbohydrate for various research and testing procedures. This 100-gram supply is ideal for experiments in food science, polymer chemistry, and microbiology. As an insoluble starch, it is commonly used in studies related to digestion, enzymatic activity, and starch hydrolysis, providing a reliable substrate for testing amylases and other carbohydrate-active enzymes. The product is frequently employed in microbiological assays, including microbial growth studies and fermentation processes, where starch serves as an energy source for specific bacteria or fungi.
Catalog No. 50-006-60870 Supplier Alkali Scientific Supplier No. TAT001CP
Potato Starch, Insoluble, is a high-purity starch used in laboratory applications requiring a stable, insoluble carbohydrate for various research and testing procedures. This 100-gram supply is ideal for experiments in food science, polymer chemistry, and microbiology. As an insoluble starch, it is commonly used in studies related to digestion, enzymatic activity, and starch hydrolysis, providing a reliable substrate for testing amylases and other carbohydrate-active enzymes. The product is frequently employed in microbiological assays, including microbial growth studies and fermentation processes, where starch serves as an energy source for specific bacteria or fungi.