Learn More
Bachem RF9 Trifluoroacetate
SDP
Supplier:  Bachem 4061350.0005
Encompass_Preferred
RF9 Trifluoroacetate, 1-Adamantanecarbonyl-Arg-Phe-NH2, 876310-60-0, C26H38N6O3, Mr 482.63, 5mg. Potent and selective neuropeptide FF (NPFF) receptor antagonist. RF9 (1-Adamantanecarbonyl-Arg-Phe-NH2) blocked the delayed and long-lasting paradoxical opioid-induced hyperalgesia and prevented the development of associated tolerance. RF9 could also efficiently block the increase in blood pressure and heart rate evoked by NPFF.
Catalog No. 50-194-6784