Learn More
Bachem (Phe2,Orn8)-Oxytocin Trifluoroacetate
SDP
Supplier:  Bachem 4030889.0025
Encompass_Preferred
(Phe2,Orn8)-Oxytocin Trifluoroacetate, H-Cys-Phe-Ile-Gln-Asn-Cys-Pro-Orn-Gly-NH2, 2480-41-3, C42H65N13O11S2, Mr 992.19, 25mg. (Phe2,Ile3,Orn8)-Vasopressin, SKF 105349. Potent vasopressor (V1) agonist with very little antidiuretic (V2) activity.
Catalog No. 50-194-6786