Learn More
Newcomer Supply Potassium Ferrocyanide 10%, Aqueous, 500 ml
SDP
Catalog No. NC9858001
Encompass
Use: Gomori Mod. Iron Stain & stock for Colloidal Iron. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
Catalog No. NC9858001 Supplier Newcomer Supply Supplier No. 13392A
Use: Gomori Mod. Iron Stain & stock for Colloidal Iron. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.