Learn More
Sigma Aldrich Fine Chemicals Biosciences Potter-Elvehjem PTfE pestl1EA
Catalog No. 502900441
Encompass
All glass construction but with a PTfE pestle and an unground tube. Rounded shape on the lower portion of the pestle. Sample is progressively ground smaller in the rounded section. Homogenate is produced as the sample is forced through the cylindrical section. Recommend for use with softer tissues.Clearance between pestle and tube is 0.004 in. to 0.006 in.
Catalog No. 50-290-0441 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P78591EA
All glass construction but with a PTfE pestle and an unground tube. Rounded shape on the lower portion of the pestle. Sample is progressively ground smaller in the rounded section. Homogenate is produced as the sample is forced through the cylindrical section. Recommend for use with softer tissues.Clearance between pestle and tube is 0.004 in. to 0.006 in.