Learn More
Electron Microscopy Sciences Potter-Elvehjem Tissue Grinder, 10 ml, 16 mmx 74 mm Mortor, 219 mm OAL
SDP
Catalog No. 501900306
Encompass_Preferred
These grinder are precisely constructed with completely interchangeable parts to allow easier cleaning or replacement of damaged mortars. Ideal for tissue homogenates and other similar applications. The Teflon pestle is for soft tissues such as brain or liver. The stainless steel rod is 0.25" (6.3 mm) diameter. The grinding chamber clearance is 0.004" to 0.006' (0.1 to 0.15 mm). This unit is autoclavable and can be used for power homogenization as with overhead stirrer.
Size: 10 ml
Mortor: 16 mmx 74 mm
OAL: 219 mm
Catalog No. 50-190-0306 Supplier Electron Microscopy Sciences Supplier No. 6479210
These grinder are precisely constructed with completely interchangeable parts to allow easier cleaning or replacement of damaged mortars. Ideal for tissue homogenates and other similar applications. The Teflon pestle is for soft tissues such as brain or liver. The stainless steel rod is 0.25" (6.3 mm) diameter. The grinding chamber clearance is 0.004" to 0.006' (0.1 to 0.15 mm). This unit is autoclavable and can be used for power homogenization as with overhead stirrer.
Size: 10 ml
Mortor: 16 mmx 74 mm
OAL: 219 mm