Learn More
Electron Microscopy Sciences Potter-Elvehjem Tissue Grinder Pestle, 2 ml, 11 mm x 45 mm Mortor, 203 mm OAL
SDP
Catalog No. 501900316
Encompass_Preferred
Replacement part to use in Potter-Elvehjem tissue grinder.
Size: 2 ml
Mortor: 11 mm x 45 mm
OAL: 203 mm
Catalog No. 50-190-0316 Supplier Electron Microscopy Sciences Supplier No. 6479280
Replacement part to use in Potter-Elvehjem tissue grinder.
Size: 2 ml
Mortor: 11 mm x 45 mm
OAL: 203 mm