Learn More
Sklar Instruments Potts-Smith Dressing Forceps, Sklar 12 in., Straight, Serrated, Stainless Steel. EA.
SDP
Catalog No. 501181428 Shop All Sklar Instruments Products
Encompass_Preferred
Potts-Smith Dressing Forceps, Sklar. 12 in., Straight, Serrated, Stainless Steel. Sklar Potts-Smith Dressing Forceps are most commonly used for application and changing of dressings. They may also be used for dissection of fine or single-layer tissue. Serrated tips provide a solid grip on both materials and tissue. This product is straight with serrated tips and a length of 12 inches.
Catalog No. 50-118-1428 Supplier Sklar Instruments Supplier No. 523312
Potts-Smith Dressing Forceps, Sklar. 12 in., Straight, Serrated, Stainless Steel. Sklar Potts-Smith Dressing Forceps are most commonly used for application and changing of dressings. They may also be used for dissection of fine or single-layer tissue. Serrated tips provide a solid grip on both materials and tissue. This product is straight with serrated tips and a length of 12 inches.