Learn More
Sklar Instruments Forceps, Potts-Smith Dressing Forceps, Sklar 8-1/4 in., Straight, Serrated, Very Delicate, Stainless Steel.
SDP
Catalog No. 501109634 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Potts-Smith Dressing Forceps, Sklar, 8-1/4 in., Straight, Serrated, Very Delicate, Stainless Steel. Sklar Potts-Smith Dressing Forceps are most commonly used for application and changing of dressings. They may also be used for dissection of fine or single-layer tissue. Serrated tips provide a solid grip on both materials and tissue. This product is straight with delicate, serrated tips and a length of 8-1/4 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-110-9634 Supplier Sklar Instruments Supplier No. 523282
Forceps, Potts-Smith Dressing Forceps, Sklar, 8-1/4 in., Straight, Serrated, Very Delicate, Stainless Steel. Sklar Potts-Smith Dressing Forceps are most commonly used for application and changing of dressings. They may also be used for dissection of fine or single-layer tissue. Serrated tips provide a solid grip on both materials and tissue. This product is straight with delicate, serrated tips and a length of 8-1/4 inches. Lifetime warranty against all manufacturer defects.