Learn More
Sklar Instruments Forceps, Potts-Smith Tissue Forceps, Sklar 7 in., 1x2 Teeth, Straight, Very Delicate, Stainless Steel, EA.
SDP
Catalog No. 501109636 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Potts-Smith Tissue Forceps, Sklar, 1x2 Teeth, Straight, Very Delicate, Stainless Steel, EA. Sklar Potts-Smith Forceps are medium weight thumb forceps most commonly used for grasping fine or delicate tissue. They are narrow forceps that feature 1x2 teeth to securely grasp the tissue, but also have a stop peg to prevent an overly harsh grasp that may crush the tissue. These forceps are most commonly used on fine or single layer tissues. This product is straight with very delicate 1x2 teeth and a length of 7 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-110-9636 Supplier Sklar Instruments Supplier No. 523370
Forceps, Potts-Smith Tissue Forceps, Sklar, 1x2 Teeth, Straight, Very Delicate, Stainless Steel, EA. Sklar Potts-Smith Forceps are medium weight thumb forceps most commonly used for grasping fine or delicate tissue. They are narrow forceps that feature 1x2 teeth to securely grasp the tissue, but also have a stop peg to prevent an overly harsh grasp that may crush the tissue. These forceps are most commonly used on fine or single layer tissues. This product is straight with very delicate 1x2 teeth and a length of 7 inches. Lifetime warranty against all manufacturer defects.