Learn More
Aviva Systems Biology Corp POU1F1 Antibody - C-terminal region (ARP31419_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31419_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POU1F1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human POU1F1. Alternate names: PIT1, CPHD1, GHF-1, Pit-1, POU1F1aApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-52826