Learn More
Aviva Systems Biology Corp POU2F1 Antibody - C-terminal region (ARP37619_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37619_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against POU2F1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of mouse POU2F1. Alternate names: Oct1, Otf1, NF-A1, Oct-1, Otf-1, 2810482H01RikApplicationWB. Species reactivity: Mouse
Catalog No. 50-014-56385