Learn More
Aviva Systems Biology Corp POU2F1 Antibody - N-terminal region (P100783_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100783_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POU2F1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human POU2F1. Alternate names: OCT1, OTF1, oct-1BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53802