Learn More
Aviva Systems Biology Corp POU2F3 Antibody - N-terminal region (ARP32537_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32537_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against POU2F3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human POU2F3. Alternate names: PLA1, OCT11, PLA-1, Epoc-1, OCT-11, OTF-11, Skn-1aApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-54642