Learn More
Aviva Systems Biology Corp POU3F1 Antibody - N-terminal region (ARP33061_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33061_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against POU3F1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human POU3F1. Alternate names: OCT6, OTF6, SCIPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52888