Learn More
Aviva Systems Biology Corp Pou3f1 Antibody - N-terminal region (ARP58050_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58050_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pou3f1. It was validated on Western Blot. . Alternate names: Oct-6, Otf6, Scip, Test1, Tst-1, Tst1, Oct6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-286-431