Learn More
Aviva Systems Biology Corp POU3F2 Antibody - middle region (ARP31218_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31218_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POU3F2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human POU3F2. Alternate names: BRN2, OCT7, OTF7, POUF3, OTF-7, brn-2, oct-7ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-54186