Learn More
Aviva Systems Biology Corp POU3F3 Antibody - N-terminal region (ARP33062_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33062_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POU3F3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human POU3F3. Alternate names: BRN1, OTF8ApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-54866